3-Bromo-5-nitrobenzenesulfonyl chloride 95%

Supplier: APOLLO SCIENTIFIC

OR400678-1G OR400678-250MG OR400678-5G
77409-480EA 447.26 USD
77409-480 77409-482 77409-484
3-Bromo-5-nitrobenzenesulfonyl chloride 95%
3-Bromo-5-nitrobenzenesulfonyl chloride
Formula: C₆H₃BrClNO₄S
MW: 300.514 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16679614
CAS Number: 1211517-87-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR