(6-Methoxybenzo[b]furan-3-yl)acetic acid 97%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 2-(6-Methoxybenzofuran-3-yl)acetic acid, 3-(Carboxymethyl)-6-methoxybenzo[b]furan

OR25758-10G OR25758-1G
77407-642EA 381.04 USD
77407-642 77407-644
(6-Methoxybenzo[b]furan-3-yl)acetic acid 97%
(6-Methoxybenzo[b]furan-3-yl)acetic acid
Formula: C₁₁H₁₀O₄
MW: 206.195 g/mol
Melting Pt: 124…127 °C
Storage Temperature: Ambient
MDL Number: MFCD02677734
CAS Number: 69716-05-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR