1-Adamantaneacetic acid 98%

Supplier: APOLLO SCIENTIFIC

OR22506-10G OR22506-25G OR22506-5G
77406-900EA 24.46 USD
77406-900 77406-902 77406-904
1-Adamantaneacetic acid 98%
1-Adamantaneacetic acid
Formula: C₁₂H₁₈O₂
Melting Pt: 137 °C
Storage Temperature: Ambient
MDL Number: MFCD00074728
CAS Number: 4942-47-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR