Methyl-3-bromobenzoate

Supplier: APOLLO SCIENTIFIC
Warning

OR2197-100G OR2197-25G
77406-756EA 104.94 USD
77406-756 77406-758
Methyl-3-bromobenzoate
Methyl-3-bromobenzoate
Formula: C₈H₇BrO₂
Boiling Pt: 127…128 °C
Melting Pt: 31…33 °C
Storage Temperature: Ambient
MDL Number: MFCD00017777
CAS Number: 618-89-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR