3-(2-Chlorophenyl)morpholine hydrochloride

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 1-Chloro-2-(morpholin-3-yl)benzene hydrochloride

OR12857-100MG OR12857-250MG
77404-446EA 358.62 USD
77404-446 77404-448
3-(2-Chlorophenyl)morpholine hydrochloride
3-(2-Chlorophenyl)morpholine hydrochloride
Formula: C₁₀H₁₂ClNO·ClH
MW: 234.122 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03840094
CAS Number: 1172507-31-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR