4-Pyridineboronic acid 97%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 4-Pyridinylboronic acid, Pyridine-4-boronic acid, Pyridin-4-ylboronic acid

OR0727-100G OR0727-25G OR0727-5G
77402-204EA 126.34 USD
77402-204 77402-206 77402-208
4-Pyridineboronic acid 97%
4-Pyridineboronic acid
Formula: C₅H₆BNO₂
MW: 122.92 g/mol (anhy)
Melting Pt: >300 °C
Storage Temperature: Freezer
MDL Number: MFCD01074545
CAS Number: 1692-15-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR