4-(Bromomethyl)-3-fluorobenzonitrile 97%

Supplier: APOLLO SCIENTIFIC
Danger

PC9915-1G PC9915-25G PC9915-5G
77418-778EA 21.26 USD
77418-778 77418-780 77418-782
4-(Bromomethyl)-3-fluorobenzonitrile 97%
4-(Bromomethyl)-3-fluorobenzonitrile
Formula: C₈H₅BrFN
Melting Pt: 75…77 °C
Storage Temperature: Ambient
MDL Number: MFCD07368341
CAS Number: 105942-09-4
UN: 3439
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR