α-Chloro-2,3,4,5-tetrafluorotoulene

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 2,3,4,5-Tetrafluorobenzyl chloride

PC9598-1G PC9598-5G
77418-686EA 38.72 USD
77418-686 77418-688
α-Chloro-2,3,4,5-tetrafluorotoulene
α-Chloro-2,3,4,5-tetrafluorotoulene
Formula: C₇H₃ClF₄
Boiling Pt: 125 °C/7 mmHg
Storage Temperature: Refrigerator
CAS Number: 21622-18-4
UN: 3265
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR