Ethyl pyrazine-2-carboxylate

Supplier: APOLLO SCIENTIFIC

OR110404-1G OR110404-250MG OR110404-5G
77403-314EA 35.68 USD
77403-314 77403-316 77403-318
Ethyl pyrazine-2-carboxylate
Ethyl pyrazine-2-carboxylate
Formula: C₇H₈N₂O₂
Boiling Pt: 120 °C/6 mmHg
Melting Pt: 49…52 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00792887
CAS Number: 6924-68-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR