Perfluorobutane-1-sulfonic acid 97%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: Perfluoro-1-butanesulphonic acid, Nonafluoro-1-butanesulphonic acid

PC9086-25G PC9086-5G
77418-546EA 135.5 USD
77418-546 77418-548
Perfluorobutane-1-sulfonic acid 97%
Perfluorobutane-1-sulphonic acid

Nonaflates are slightly more reactive towards nucleophilic displacements than triflates.

Formula: C₄HF₉O₃S
Boiling Pt: 76…84 °C/0.1 mmHg
Density: 1.811 g/cm³ (25 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD01320794
CAS Number: 375-73-5
EINECS: 206-793-1
UN: 2586
ADR: 8 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR