(1R)-1-(3-Fluorophenyl)ethylamine 97%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: (R)-1-(3-Fluorophenyl)ethanamine, (R)-3-Fluoro-α-methylbenzylamine

PC9079-1G PC9079-250MG
77418-538EA 103.92 USD
77418-538 77418-540
(1R)-1-(3-Fluorophenyl)ethylamine 97%
(1R)-1-(3-Fluorophenyl)ethylamine
Formula: C₈H₁₀FN
MW: 139.17 g/mol
Flash Pt: 72 °C
Storage Temperature: Ambient
MDL Number: MFCD06761859
CAS Number: 761390-58-3
UN: 2735
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR