2-Bromo-3-fluoropyridine 98%

Supplier: APOLLO SCIENTIFIC
Warning

PC8846-100G PC8846-25G PC8846-5G
77418-462EA 114.1 USD
77418-462 77418-464 77418-466
2-Bromo-3-fluoropyridine 98%
2-Bromo-3-fluoropyridine
Formula: C₅H₃BrFN
Density: 1.74 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD04114128
CAS Number: 40273-45-8
UN: 2810
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR