4,4,4-Trifluorobutylamine 97%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 1-Butanamine,4,4,4-trifluoro-, 4,4,4-trifluorobutan-1-amine, 1-Amino-4,4,4-trifluorobutane

PC9728-1G PC9728-5G
77418-728EA 49.92 USD
77418-728 77418-730
4,4,4-Trifluorobutylamine 97%
4,4,4-Trifluorobutylamine
Formula: C₄H₈F₃N
MW: 127.11 g/mol
Boiling Pt: 90…94 °C
Density: 1.35 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD06213030
CAS Number: 819-46-5
UN: 2734
ADR: 8+3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR