3,5-Bis(trifluoromethyl)anisole 98%

Supplier: APOLLO SCIENTIFIC
Warning

PC9670-10G PC9670-25G PC9670-5G
77418-716EA 44.82 USD
77418-716 77418-718 77418-720
3,5-Bis(trifluoromethyl)anisole 98%
3,5-Bis(trifluoromethyl)anisole
Formula: C₉H₆F₆O
Boiling Pt: 158…160 °C
Flash Pt: 55 °C
Storage Temperature: Ambient
MDL Number: MFCD00728736
CAS Number: 349-60-0
UN: 1993
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR