2-(4-Fluorophenyl)propan-2-amine

Supplier: APOLLO SCIENTIFIC
Danger

PC8957-10G PC8957-1G PC8957-5G
77418-492EA 511.44 USD
77418-492 77418-494 77418-496
2-(4-Fluorophenyl)propan-2-amine
2-(4-Fluorophenyl)propan-2-amine
Formula: C₉H₁₂FN
Boiling Pt: 40 °C/5.25 mmHg
Flash Pt: 92 °C
Storage Temperature: Refrigerator
MDL Number: MFCD07777176
CAS Number: 17797-10-3
UN: 3259
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR