(4-Bromo-2-fluorophenyl)acetonitrile

Supplier: APOLLO SCIENTIFIC
Warning

PC8933-100G PC8933-25G PC8933-5G
77418-482EA 118.5 USD
77418-482 77418-484 77418-486
(4-Bromo-2-fluorophenyl)acetonitrile
(4-Bromo-2-fluorophenyl)acetonitrile
Formula: C₈H₅BrFN
Melting Pt: 52…53 °C
Storage Temperature: Ambient
MDL Number: MFCD07368653
CAS Number: 114897-91-5
UN: 3439
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR