5-Chloro-2-fluoro-4-methylpyridine

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 5-Chloro-2-fluoro-4-picoline

PC8385-1G PC8385-25G PC8385-5G
77418-400EA 25 USD
77418-400 77418-402 77418-404
5-Chloro-2-fluoro-4-methylpyridine
5-Chloro-2-fluoro-4-methylpyridine
Formula: C₆H₅ClFN
MW: 145.56 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04972410
CAS Number: 884494-88-6
UN: 2810
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR