(3,3,3-Trifluoropropyl)trimethoxysilane 97%
Supplier: APOLLO SCIENTIFIC
|
Warning
|
Synonyms:
Trimethoxy(3,3,3-trifluoropropyl)silane
PC7826-100G
PC7826-25G
77418-248EA
150.78
USD
77418-248
77418-250
(3,3,3-Trifluoropropyl)trimethoxysilane 97%
(3,3,3-Trifluoropropyl)trimethoxysilane
Formula:
F₃CCH₂CH₂Si(OCH₃)₃ Boiling Pt: 144 °C Density: 1.137 g/cm³ (25 °C) Flash Pt: 38 °C Storage Temperature: Ambient |
MDL Number:
MFCD00039266 CAS Number: 429-60-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...