Methyl 3-chloroisoquinoline-6-carboxylate 95%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 3-Chloro-6-(methoxycarbonyl)-2-azanaphthalene, 3-Chloro-6-(methoxycarbonyl)isoquinoline

OR300130-100MG OR300130-250MG
77408-564EA 307.68 USD
77408-564 77408-566
Methyl 3-chloroisoquinoline-6-carboxylate 95%
Methyl 3-chloroisoquinoline-6-carboxylate
Formula: C₁₁H₈ClNO₂
MW: 221.64 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22689680
CAS Number: 1416713-88-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR