3,5-Dimethyl-1H-pyrazole-4-boronic acid pinacol ester 97%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 3,5-Dimethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole, 3,5-Dimethylpyrazole-4-boronic acid pinacol ester

OR2255-1G OR2255-25G OR2255-5G
77406-906EA 89.66 USD
77406-906 77406-908 77406-910
3,5-Dimethyl-1H-pyrazole-4-boronic acid pinacol ester 97%
3,5-Dimethyl-1H-pyrazole-4-boronic acid pinacol ester
Formula: C₁₁H₁₉BN₂O₂
Melting Pt: 163…168 °C
Storage Temperature: Ambient
MDL Number: MFCD05663852
CAS Number: 857530-80-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR