1-(Piperidine-1-sulfonyl)piperazine

Supplier: APOLLO SCIENTIFIC
Warning

OR0954-1G OR0954-250MG
77402-336EA 110.04 USD
77402-336 77402-338
1-(Piperidine-1-sulfonyl)piperazine
1-(Piperidine-1-sulfonyl)piperazine
Formula: C₉H₁₉N₃O₂S
Storage Temperature: Ambient
MDL Number: MFCD04973338
CAS Number: 500587-48-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR