4-Aminothiophenol 97%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 4-Aminobenzenethiol, 4-Aminophenyl mercaptan, 4-Mercaptoaniline

OR0772-25G OR0772-500G OR0772-100G
77402-224EA 50.94 USD
77402-224 77402-226 77402-222
4-Aminothiophenol 97%
4-Aminothiophenol
Formula: C₆H₇NS
MW: 125.19 g/mol
Boiling Pt: 140…145 °C/16 mmHg
Melting Pt: 39…42 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00007880
CAS Number: 1193-02-8
UN: 3263
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR