3-(Trifluoromethoxy)cinnamic acid 96%

Supplier: APOLLO SCIENTIFIC
Danger

PC8754-100G PC8754-25G PC8754-5G
77418-444EA 174.22 USD
77418-444 77418-446 77418-448
3-(Trifluoromethoxy)cinnamic acid 96%
3-(Trifluoromethoxy)cinnamic acid
Formula: C₁₀H₇F₃O₃
Melting Pt: 92…96 °C
Storage Temperature: Ambient
MDL Number: MFCD00066337
CAS Number: 168833-80-5
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR