3-Fluoro-p-cresol

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 3-Fluoro-4-methylphenol, 1-Fluoro-3-hydroxy-6-methylbenzene

PC8221-1G PC8221-25G PC8221-5G
77418-360EA 21.26 USD
77418-360 77418-362 77418-364
3-Fluoro-p-cresol
3-Fluoro-p-cresol
Formula: C₇H₇FO
MW: 126.13 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00143164
CAS Number: 452-78-8
UN: 3265
ADR: 8 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR