α,α,α-Trifluoro-o-toluoyl chloride 98%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 2-(Trifluoromethyl)benzoyl chloride, 2-(Trifluoromethyl)benzene-1-carbonyl chloride

PC7590-100G PC7590-25G PC7590-500G
77418-014EA 54 USD
77418-014 77418-016 77418-018
α,α,α-Trifluoro-o-toluoyl chloride 98%
α,α,α-Trifluoro-o-toluoyl chloride
Formula: C₈H₄ClF₃O
Boiling Pt: 84…85 °C/16 mmHg
Melting Pt: –22 °C
Density: 1.416 g/cm³ (25 °C)
Flash Pt: 95 °C
Storage Temperature: Ambient
MDL Number: MFCD00000667
CAS Number: 312-94-7
EINECS: 206-233-6
UN: 3265
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR