Zinc trifluoromethanesulphonate 98%
Supplier: APOLLO SCIENTIFIC
|
Danger
|
Synonyms:
Zinc(II) trifluoromethanesulphonate, Zinc triflate, Zinc trifluoromethanesulphonate
PC8034-100G
PC8034-25G
PC8034-5G
77418-340EA
143.66
USD
77418-340
77418-342
77418-344
Zinc trifluoromethanesulphonate 98%
Zinc trifluoromethanesulphonate
Formula:
C₂F₆O₆S₂Zn MW: 363.53 Melting Pt: >300 °C Storage Temperature: Ambient |
MDL Number:
MFCD00013229 CAS Number: 54010-75-2 EINECS: 258-922-6 UN: 3261 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...