Trimethyloxonium tetrafluoroborate 95%

Supplier: APOLLO SCIENTIFIC
Danger

PC7865-100G PC7865-25G PC7865-5G
77418-262EA 95.76 USD
77418-262 77418-264 77418-266
Trimethyloxonium tetrafluoroborate 95%
Trimethyloxonium tetrafluoroborate

Versatile mild methylating agent.

Formula: C₃H₉BF₄O
MW: 147.91 g/mol
Melting Pt: 200 °C
Flash Pt: >110 °C
Storage Temperature: Freezer
MDL Number: MFCD00011798
CAS Number: 420-37-1
EINECS: 206-994-4
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR