2,2,2-Trifluoroethyl methacrylate 99%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: Methacrylic acid 2,2,2-trifluoroethyl ester

PC7374-100G PC7374-25G PC7374-500G
77417-840EA 39.74 USD
77417-840 77417-842 77417-844
2,2,2-Trifluoroethyl methacrylate 99%
2,2,2-Trifluoroethyl methacrylate
Formula: C₆H₇F₃O₂
MW: 168.12 g/mol
Boiling Pt: 29…30 °C/40 mmHg
Density: 1.181 g/cm³ (25 °C)
Flash Pt: 16 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00013576
CAS Number: 352-87-4
EINECS: 206-525-3
UN: 3272
ADR: 3 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR