1-Fluoropropan-2-ol 97%

Supplier: APOLLO SCIENTIFIC
Warning

PC7229-1G PC7229-5G
77417-686EA 172.18 USD
77417-686 77417-688
1-Fluoropropan-2-ol 97%
1-Fluoropropan-2-ol
Formula: C₃H₇FO
Boiling Pt: 107…108 °C
Density: 1.021 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00039620
CAS Number: 430-50-2
UN: 1987
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR