Trifluoroacetic anhydride 99%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: Trifluoracetic anhydride, TFAA

PC7170-1KG PC7170-500G
77417-676EA 103.48 USD
77417-676 77417-678
Trifluoroacetic anhydride 99%
Trifluoroacetic anhydride

Useful trifluoroacetylating agent for the protectionof alcohols & amines.

Formula: C₄F₆O₃
MW: 210.03 g/mol
Boiling Pt: 39…40 °C
Density: 1.511 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000416
CAS Number: 407-25-0
EINECS: 206-982-9
UN: 3265
ADR: 8,I

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR