1,2,3,4-Tetrafluorobenzene 98%
Supplier: APOLLO SCIENTIFIC
|
Danger
|
PC6710-100G
PC6710-25G
PC6710-5G
77417-502EA
296.48
USD
77417-502
77417-504
77417-506
1,2,3,4-Tetrafluorobenzene 98%
1,2,3,4-Tetrafluorobenzene
Formula:
C₆H₂F₄ Boiling Pt: 94…96 °C Melting Pt: –42 °C Density: 1.4 g/cm³ (25 °C) Flash Pt: 20 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000285 CAS Number: 551-62-2 EINECS: 208-997-6 UN: 1993 ADR: 3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...