(3,3,3-Trifluoropropyl)dichloromethylsilane 97%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: Dichloro(methyl)(3,3,3-trifluoropropyl)silane, (3,3,3-Trifluoropropyl)methyldichlorosilane

PC7820FB-100G PC7820FB-25G
77418-240EA 198.66 USD
77418-240 77418-242
(3,3,3-Trifluoropropyl)dichloromethylsilane 97%
(3,3,3-Trifluoropropyl)dichloromethylsilane
Formula: C₄H₇Cl₂F₃Si
MW: 211.09
Boiling Pt: 121…122 °C
Density: 1.261 g/cm³ (25 °C)
Flash Pt: 15 °C
Storage Temperature: Ambient
MDL Number: MFCD00039294
CAS Number: 675-62-7
EINECS: 211-623-4
UN: 2985
ADR: 3+8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR