4-(Trifluoromethyl)phenyl isothiocyanate 98%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 1-Isocyanato-4-(trifluoromethyl)benzene, α,α,α-Trifluoro-p-tolyl isothiocyanate, p-Trifluoromethylphenylisothiocyanate

PC7741-1G PC7741-5G
77418-158EA 44.82 USD
77418-158 77418-160
4-(Trifluoromethyl)phenyl isothiocyanate 98%
4-(Trifluoromethyl)phenyl isothiocyanate
Formula: C₈H₄F₃NS
MW: 203.19 g/mol
Boiling Pt: 207 °C
Melting Pt: 39…43 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00039645
CAS Number: 1645-65-4
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR