3-(Trifluoromethyl)benzyl mercaptan, Technical Grade
Supplier: APOLLO SCIENTIFIC
|
Warning
|
Synonyms:
3-(Trifluoromethyl)benzylthiol
PC7634-10G
PC7634-1G
PC7634-25G
77418-070EA
96.78
USD
77418-070
77418-072
77418-074
3-(Trifluoromethyl)benzyl mercaptan, Technical Grade
3-(Trifluoromethyl)benzyl mercaptan
Formula:
C₈H₇F₃S MW: 192.2 g/mol Boiling Pt: 75…77 °C/5 mmHg Storage Temperature: Ambient |
MDL Number:
MFCD00042435 CAS Number: 25697-55-6 UN: 2810 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...