4-(Trifluoromethoxy)phenylacetonitrile 98%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 1-(Cyanmethyl)-4-(trifluormethoxy)benzol, 4-(TrifluoroMethoxy)phenylacetonitrile, 4-(Trifluoromethoxy)benzyl cyanide

PC7440H-100G PC7440H-25G PC7440H-5G
77417-978EA 121.24 USD
77417-978 77417-980 77417-982
4-(Trifluoromethoxy)phenylacetonitrile 98%
4-(Trifluoromethoxy)phenylacetonitrile
Formula: C₉H₆F₃NO
MW: 201.15 g/mol
Boiling Pt: 96 °C/3.75 mmHg
Melting Pt: 28…33 °C
Density: 1.28 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00061240
CAS Number: 49561-96-8
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR