2-(Trifluoromethoxy)benzoyl chloride 97%

Supplier: APOLLO SCIENTIFIC
Danger

PC7438L-100G PC7438L-25G PC7438L-5G
77417-896EA 99.84 USD
77417-896 77417-898 77417-900
2-(Trifluoromethoxy)benzoyl chloride 97%
2-(Trifluoromethoxy)benzoyl chloride
Formula: C₈H₄ClF₃O₂
Boiling Pt: 94…96 °C/15 mmHg
Density: 1.435 g/cm³ (25 °C)
Flash Pt: 102 °C
Storage Temperature: Ambient
MDL Number: MFCD00052330
CAS Number: 162046-61-9
UN: 3265
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR