2,2,3,3-Tetrafluoropropionic acid 97%

Supplier: APOLLO SCIENTIFIC
Danger

PC6866E-25G PC6866E-5G
77417-574EA 170.54 USD
77417-574 77417-576
2,2,3,3-Tetrafluoropropionic acid 97%
2,2,3,3-Tetrafluoropropionic acid
Formula: C₃H₂F₄O₂
MW: 146.04 g/mol
Boiling Pt: 133…134 °C
Density: 1.563 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00054686
CAS Number: 756-09-2
EINECS: 212-049-7
UN: 3265
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR