Sodium fluoride 99.99%

Supplier: APOLLO SCIENTIFIC
Danger

PC6466B-1G PC6466B-25G PC6466B-5G
77417-404EA 21.26 USD
77417-404 77417-406 77417-408
Sodium fluoride 99.99%
Sodium fluoride
Formula: NaF
MW: 41.99 g/mol
Melting Pt: 988 °C
Density: 2.78 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003524
CAS Number: 7681-49-4
EINECS: 231-667-8
UN: 1690
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR