1,1,1-Trifluoro-3-phenylacetone 98%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 3-Phenyl-1,1,1-trifluoropropan-2-one, 1,1,1-Trifluoro-3-phenyl-2-propanone, (3,3,3-Trifluor-2-propanoyl)benzene, 1,1,1-Trifluoro-3-phenylpropan-2-one

PC6229M-1G PC6229M-25G PC6229M-5G
77417-328EA 41.78 USD
77417-328 77417-330 77417-332
1,1,1-Trifluoro-3-phenylacetone 98%
1,1,1-Trifluoro-3-phenylacetone
Formula: C₉H₇F₃O
MW: 188.15 g/mol
Boiling Pt: 51…52 °C/2 mmHg
Density: 1.22 g/cm³ (25 °C)
Flash Pt: 33 °C
Storage Temperature: Ambient
MDL Number: MFCD00068171
CAS Number: 350-92-5
UN: 1224
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR