8-Quinolinesulfonyl chloride

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: Quinoline-8-sulphonyl chloride

OR23069-100G OR23069-25G OR23069-5G
77407-066EA 268.96 USD
77407-066 77407-068 77407-070
8-Quinolinesulfonyl chloride
8-Quinolinesulphonyl chloride
Formula: C₉H₆ClNO₂S
MW: 227.67 g/mol
Melting Pt: 126…129 °C
Storage Temperature: Ambient
MDL Number: MFCD00006808
CAS Number: 18704-37-5
EINECS: 242-515-5
UN: 3261
ADR: 8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR