3,3,3-Trifluoro-1-propanol 97%

Supplier: APOLLO SCIENTIFIC
Warning

PC7819R-1G PC7819R-25G PC7819R-5G
77418-228EA 21.32 USD
77418-228 77418-230 77418-232
3,3,3-Trifluoro-1-propanol 97%
3,3,3-Trifluoro-1-propanol
Formula: C₃H₅F₃O
Boiling Pt: 100…101 °C
Density: 1.294 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00190649
CAS Number: 2240-88-2
UN: 1987
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR