3,4,5-Trifluorophenol 95%

Supplier: APOLLO SCIENTIFIC
Danger

PC7818-100G PC7818-25G PC7818-5G
77418-216EA 135.5 USD
77418-216 77418-218 77418-220
3,4,5-Trifluorophenol 95%
3,4,5-Trifluorophenol
Formula: C₆H₃F₃O
MW: 148.08 g/mol
Boiling Pt: 177 °C
Melting Pt: 52…55 °C
Flash Pt: 87 °C
Storage Temperature: Ambient
MDL Number: MFCD00042427
CAS Number: 99627-05-1
EINECS: 436-120-9
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR