2,4,5-Trifluorobenzoyl chloride 99%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 2,4,5-trifluorobenzoylchloride

PC7285G-100G PC7285G-25G PC7285G-5G
77417-722EA 174.3 USD
77417-722 77417-724 77417-726
2,4,5-Trifluorobenzoyl chloride 99%
2,4,5-Trifluorobenzoyl chloride
Formula: C₇H₂ClF₃O
MW: 194.54 g/mol
Boiling Pt: 85 °C/17 mmHg
Density: 1.52 g/cm³ (25 °C)
Flash Pt: 86 °C
Storage Temperature: Ambient
MDL Number: MFCD00061204
CAS Number: 88419-56-1
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR