2,4,6-Trifluorobenzonitrile 98%

Supplier: APOLLO SCIENTIFIC
Warning

PC7283R-100G PC7283R-25G PC7283R-5G
77417-714EA 146.7 USD
77417-714 77417-716 77417-718
2,4,6-Trifluorobenzonitrile 98%
2,4,6-Trifluorobenzonitrile
Formula: C₇H₂F₃N
MW: 157.09 g/mol
Boiling Pt: 179 °C
Melting Pt: 55…63 °C
Flash Pt: 92 °C
Storage Temperature: Ambient
MDL Number: MFCD00042399
CAS Number: 96606-37-0
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR