Tetrafluorophthalonitrile 97%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 3,4,5,6-Tetrafluorophthalonitrile, 1,2-Dicyano-3,4,5,6-tetrafluorobenzene

PC6860-100G PC6860-10G PC6860-25G
77417-562EA 104.94 USD
77417-562 77417-564 77417-566
Tetrafluorophthalonitrile 97%
Tetrafluorophthalonitrile
Formula: C₈F₄N₂
MW: 200.1 g/mol
Melting Pt: 81…86 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00001774
CAS Number: 1835-65-0
EINECS: 217-400-8
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR