2-Hydrazino-4-(trifluoromethyl)pyrimidine 97%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 2-Hydrazinyl-4-(trifluoromethyl)pyrimidine, [4-(Trifluoromethyl)pyrimidin-2-yl]hydrazine

PC6431-1G PC6431-5G
77417-382EA 20.32 USD
77417-382 77417-384
2-Hydrazino-4-(trifluoromethyl)pyrimidine 97%
2-Hydrazino-4-(trifluoromethyl)pyrimidine
Formula: C₅H₅F₃N₄
MW: 178.118 g/mol
Melting Pt: 99…101 °C
Storage Temperature: Ambient
MDL Number: MFCD00115112
CAS Number: 197305-97-8
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR