5H-Octafluoropentanoic acid 97%

Supplier: APOLLO SCIENTIFIC
Danger

PC6185-5G PC6185-1G
77417-302EA 32 USD
77417-302 77417-304
5H-Octafluoropentanoic acid 97%
5H-Octafluoropentanoic acid
Formula: C₅H₂F₈O₂
Boiling Pt: 165…166 °C
Density: 1.725 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00054690
CAS Number: 376-72-7
EINECS: 206-812-3
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR