2-Fluoro-4-methoxybenzoyl chloride 95%

Supplier: APOLLO SCIENTIFIC

PC9192-1G PC9192-5G
77418-610EA 168.1 USD
77418-610 77418-612
2-Fluoro-4-methoxybenzoyl chloride 95%
2-Fluoro-4-methoxybenzoyl chloride
Formula: C₈H₆ClFO₂
Melting Pt: 40…42 °C
Storage Temperature: Ambient
MDL Number: MFCD08146649
CAS Number: 321-24-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR