3-Fluoroprop-1-yl methanesulfonate 98%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 3-Fluoropropyl methanesulphonate, 3-Fluoroprop-1-yl mesylate, 3-Fluoroprop-1-yl methanesulphonate

PC9175-1G PC9175-250MG PC9175-5G
77418-602EA 96.78 USD
77418-602 77418-604 77418-606
3-Fluoroprop-1-yl methanesulfonate 98%
3-Fluoroprop-1-yl methanesulfonate
Formula: C₄H₉FO₃S
MW: 156.176 g/mol
Boiling Pt: 116 °C (15 mmHg)
Flash Pt: 109 °C
Storage Temperature: Ambient
MDL Number: MFCD01715281
CAS Number: 372-04-3
UN: 2810
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR