2-Bromo-4-fluoro-6-(trifluoromethyl)aniline 98%

Supplier: APOLLO SCIENTIFIC

Synonyms: 6-Bromo-α,α,α,4-tetrafluoro-o-toluidine

PC906319-100G PC906319-25G PC906319-5G
77418-532EA 399.38 USD
77418-532 77418-534 77418-536
2-Bromo-4-fluoro-6-(trifluoromethyl)aniline 98%
2-Bromo-4-fluoro-6-(trifluoromethyl)aniline
Formula: C₇H₄BrF₄N
Storage Temperature: Ambient
MDL Number: MFCD04973756
CAS Number: 875664-27-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR